Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorothiophenol, 97%, ACROS Organics™

Click to view available options
Quantity:
1mL
5mL
25mL
Packaging:
Glass bottle
771-62-0
C6HF5S
200.13
MFCD00004828
UVAMFBJPMUMURT-UHFFFAOYSA-N
pentafluorothiophenol,pentafluorobenzenethiol,benzenethiol, pentafluoro,pentafluorophenyl mercaptan,2,3,4,5,6-pentafluorothiophenol,2,3,4,5,6-pentafluorobenzene-1-thiol,pentafluorobenzene thiol,pentafluorobenzene-1-thiol,2,3,4,5,6-pentafluorothiphenol
13042
2,3,4,5,6-pentafluorobenzenethiol
C1(=C(C(=C(C(=C1F)F)S)F)F)F
771-62-0
C6HF5S
200.13
MFCD00004828
UVAMFBJPMUMURT-UHFFFAOYSA-N
pentafluorothiophenol,pentafluorobenzenethiol,benzenethiol, pentafluoro,pentafluorophenyl mercaptan,2,3,4,5,6-pentafluorothiophenol,2,3,4,5,6-pentafluorobenzene-1-thiol,pentafluorobenzene thiol,pentafluorobenzene-1-thiol,2,3,4,5,6-pentafluorothiphenol
13042
2,3,4,5,6-pentafluorobenzenethiol
C1(=C(C(=C(C(=C1F)F)S)F)F)F
Melting Point -24°C
Flash Point 51°C
Boiling Point 143°C
Refractive Index 1.4630 to 1.465
Specific Gravity 1.5
Quantity 25mL
Infrared Spectrum Authentic
Density 1.5000g/mL
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula C6F5SH
Formula Weight 200.13
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Pentafluorothiophenol
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger