Alfa Aesar™ Pentakis(diethylamino)niobium(V), 99.9% (metals basis), mixture of complexes

Nitride film precursor

Manufacturer: Alfa Aesar™ 40445ME

Quantity 500mg

 View more versions of this product

Catalog No. AA40445ME

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 25169-05-5
    Linear Formula Nb[N(C2H5)2]5
    Formula Weight 453.56
    MDL Number MFCD00798536
    UN Number UN3267
    Synonym Niobium diethylamide
    Chemical Name or Material Pentakis(diethylamino)niobium(V), Mixture of complexes

    Specifications

    Formula Weight 453.56
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    Physical Form Liquid
    MDL Number MFCD00798536
    CAS 25169-05-5
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Odor Amine-like
    Boiling Point 120°C (0.1mmHg)
    Color Dark
    UN Number UN3267
    Synonym Niobium diethylamide
    Sensitivity Moisture Sensitive
    Quantity 500mg
    Linear Formula Nb[N(C2H5)2]5
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Chemical Name or Material Pentakis(diethylamino)niobium(V), Mixture of complexes
    Assay 99.9%, (metals basis)

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P402
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit