Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pentakis(diethylamino)niobium(V), 99.9% (metals basis), mixture of complexes

Nitride film precursor

Manufacturer:  Alfa Aesar™ 4044527

Quantity 2.5g

 View more versions of this product

Catalog No. AA4044527


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

25169-05-5
453.56
UN3267
Pentakis(diethylamino)niobium(V), Mixture of complexes
Nb[N(C2H5)2]5
MFCD00798536
Niobium diethylamide

Specifications

Amine-like
120°C (0.1mmHg)
2.5g
Liquid
453.56
UN3267
Pentakis(diethylamino)niobium(V), Mixture of complexes
Moisture Sensitive
Dark
25169-05-5
Nb[N(C2H5)2]5
MFCD00798536
Niobium diethylamide
99.9%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit