Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentamethylbenzaldehyde, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
17432-38-1
C12H16O
176.259
MFCD00015560
RWOZGGOKRKSHKN-UHFFFAOYSA-N
pentamethylbenzaldehyde,pubchem16352,acmc-1busy,3,4-dibenzyloxyphenethylamine hydroxide
263712
2,3,4,5,6-pentamethylbenzaldehyde
CC1=C(C(=C(C(=C1C)C)C=O)C)C
17432-38-1
C12H16O
176.259
MFCD00015560
RWOZGGOKRKSHKN-UHFFFAOYSA-N
pentamethylbenzaldehyde,pubchem16352,acmc-1busy,3,4-dibenzyloxyphenethylamine hydroxide
263712
2,3,4,5,6-pentamethylbenzaldehyde
CC1=C(C(=C(C(=C1C)C)C=O)C)C
Quantity 1g
Infrared Spectrum Authentic
Color Yellow to Beige
Boiling Point 144.0°C (6.0 mmHg)
Packaging Glass bottle
Melting Point 146.0°C to 148.0°C
Assay Percent Range 96%
Linear Formula (CH3)5C6C(O)H
Formula Weight 176.26
Physical Form Crystalline Powder, Crystals or Flakes
Percent Purity 96%
Chemical Name or Material Pentamethylbenzaldehyde, 96%
Show More Show Less