Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pentamethylcyclopentadienyltitanium trichloride

Click to view available options
Quantity:
1g
5g
12129-06-5
MFCD00070434
12129-06-5
MFCD00070434
Color Red
Quantity 5g
Sensitivity Moisture sensitive
Melting Point 200°C (decomposition)
UN Number UN3261
Solubility Information Reacts with water
Formula Weight 289.47
Physical Form Crystalline
Chemical Name or Material Pentamethylcyclopentadienyltitanium trichloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen