Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

109-66-0
C5H12
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
8003
CHEBI:37830
pentane
CCCCC

High purity solvents for use in analytical and preparative separations

    For use in liquid chromatography (HPLC & UHPLC) & spectrophotometry
Quantity 4L
Flash Point −40.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.63kg
Formula Weight 72.15
Chemical Name or Material Pentane
Grade HPLC Grade