Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane, SpectroSolv (TM), 97%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
109-66-0
109-66-0
CAS Max % 1
Percent Purity 0.97
Grade High Purity Solvent
Packaging Amber Glass Bottle
Residue after Evaporation 5 ppm
Water 0.0002
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.