Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentane, ULTRA RESI-ANALYZED™, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
109-66-0
C5H12
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish
8003
CHEBI:37830
pentane
CCCCC
109-66-0
C5H12
72.151
OFBQJSOFQDEBGM-UHFFFAOYSA-N
n-pentane,pentan,skellysolve a,pentanen,pentani,amyl hydride,tetrafume,tetrakil,tetraspot,pentan polish
8003
CHEBI:37830
pentane
CCCCC

High purity solvents and salts developed for organic residue extraction/concentration procedures

    For use in Gas Chromatography (GC), Organic Residue Analysis; For Laboratory, Research, or Manufacturing Use
Flash Point −40.00°C(-40°F)
Packaging Narrow Mouth Amber Glass Bottle
Quantity 1L
Specific Gravity 1L = 0.63kg
Formula Weight 72.15
Grade ULTRA RESI-ANALYZED™
Chemical Name or Material Pentane