Alfa Aesar™ Pentanol, mixture of isomers, 99% (sum of isomers)

Manufacturer: Alfa Aesar™ H333910J

Quantity 5000mL

 View more versions of this product

Catalog No. AAH333910J

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 94624-12-1
    Molecular Formula C5H12O
    Formula Weight 88.15
    MDL Number MFCD00081734
    UN Number UN1987
    Synonym Amyl alcohol, mixed isomers
    Chemical Name or Material Pentanol, mixture of isomers

    Specifications

    Formula Weight 88.15
    Boiling Point 128° to 137°C
    Molecular Formula C5H12O
    Quantity 5000mL
    Flash Point 45°C (113°F)
    MDL Number MFCD00081734
    UN Number UN1987
    Synonym Amyl alcohol, mixed isomers
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H332-H315-H335
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 94624-12-1
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Chemical Name or Material Pentanol, mixture of isomers
    Assay 99%

    GHS H Statement
    H226-H332-H315-H335
    Flammable liquid and vapor.
    Harmful if inhaled.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit