Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Pentaphenylcyclopentadiene, 99%

Manufacturer:  Alfa Aesar™ H27223MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH27223MD


  Check Availability
Add to cart

Specifications

Description

Applications
1,2,3,4,5-Pentaphenyl-1,3-cyclopentadiene (PPCP) was used as dopant for poly(3-alkylthiophene) diodes to enhance their electroluminescence efficiency, in the preparation of monodisperse and highly fluorescent silica nanoparticles.

Solubility
Soluble in pyridine 5 mg/ml.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

2519-10-0
446.59
Pentaphenylcyclopentadiene
C35H26
MFCD09909572

Specifications

2519-10-0
C35H26
446.59
Pentaphenylcyclopentadiene
254° to 257°C
250mg
MFCD09909572
99%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit