Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentoxyresorufin, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
87687-03-4
C17H17NO3
283.327
ZPSOKQFFOYYPKC-UHFFFAOYSA-N
pentoxyresorufin,resorufin pentyl ether,7-pentoxyresorufin,7-pentyloxy-3h-phenoxazin-3-one,o-pentylresorufin,7-pentoxyphenoxazone,3h-phenoxazin-3-one, 7-pentyloxy,o7-pentylresorufin,7-pentyloxy-3-phenoxazone
107683
7-pentoxyphenoxazin-3-one
CCCCCOC1=CC2=C(C=C1)N=C3C=CC(=O)C=C3O2
This item is not returnable. View return policy
87687-03-4
C17H17NO3
283.327
ZPSOKQFFOYYPKC-UHFFFAOYSA-N
pentoxyresorufin,resorufin pentyl ether,7-pentoxyresorufin,7-pentyloxy-3h-phenoxazin-3-one,o-pentylresorufin,7-pentoxyphenoxazone,3h-phenoxazin-3-one, 7-pentyloxy,o7-pentylresorufin,7-pentyloxy-3-phenoxazone
107683
7-pentoxyphenoxazin-3-one
CCCCCOC1=CC2=C(C=C1)N=C3C=CC(=O)C=C3O2

Fluorometric cytochrome P450 substrate

Fluorometric cytochrome P450 substrate.
Quantity 10mg
Formula Weight 283.32
Percent Purity >98%
Chemical Name or Material Pentoxyresorufin

Recommended Storage : Store at -20°C