Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pentraxin 2/SAP, Mouse anti-Human, DyLight 594, Clone: APCS/3240, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279848DL594
Description
Pentraxin 2/SAP Monoclonal antibody specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Pentraxin 2/SAP | |
APCS/3240 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Conjugated, Purified | |
APCS | |
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 594 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title