Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Pentraxin 3/TSG-14 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
$299.26 - $642.50
Specifications
Target | Pentraxin 3/TSG-14 |
---|---|
Conjugate | HRP |
Target Species | Human |
Gene ID (Entrez) | 5806 |
Gene Symbol | PTX3 |
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Pentraxin 3/TSG-14.Specifications
Pentraxin 3/TSG-14 | |
Human | |
PTX3 |
HRP | |
5806 | |
Rabbit/Mouse |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title