Learn More
Aviva Systems Biology Corp PEPD Antibody - middle region (ARP56088_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP56088_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PEPD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PEPD. Alternate names: MGC10905, PROLIDASEApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61713