Learn More
Affymetrix/USB™ Pepsin from Porcine Stomach
Derived from Porcine stomach
Manufacturer:  Affymetrix/USB™ J2001014
 View more versions of this product
Catalog No. AAJ2001014
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.
Specifications
Specifications
Lyophilized powder
25g
EC 3.4.23.1,Pepsin A
Pepsin
9001-75-6