Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Affymetrix/USB™ Pepsin from Porcine Stomach

Derived from Porcine stomach

$751.50 - $924.00

Specifications

Synonym EC 3.4.23.1,Pepsin A
Format Lyophilized powder
CAS 9001-75-6
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ2001014 View Documents Affymetrix/USB™
J2001014
25g Each for $924.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ20015A1 View Documents Affymetrix/USB™
J20015A1
1kg Each for $751.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • One unit causes an increase in absorbance at 280nm of 0.001/min. at 37°C at pH 2.0.
Specifications

Specifications

EC 3.4.23.1,Pepsin A
Lyophilized powder
9001-75-6
Safety and Handling

Safety and Handling

EINECSNumber : 232-629-3

Recommended Storage : Store at -20°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit