Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen A Antibody (3D1C6), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306077MFV450
Description
Pepsinogen A Monoclonal antibody specifically detects Pepsinogen A in Human samples. It is validated for ELISASpecifications
Pepsinogen A | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
643847 | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
3D1C6 | |
ELISA | |
EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A (Accession#: NP_001073276.1; Met 1-Ala 388). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction