Learn More
Pepsinogen A Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP300065JF549
Catalog No. NB064201
Add to cart
Description
Description
Pepsinogen A Polyclonal specifically detects Pepsinogen A in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Pepsinogen A
Janelia Fluor 549
50mM Sodium Borate
EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5
Produced in rabbits immunized with purified, recombinant Pepsinogen A (P00790; Met1-Ala388).
Affinity purified
RUO
Primary
643847
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Store at 4°C in the dark.
Polyclonal
Human