Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pepsinogen C/PGC/Progastricsin Antibody (90/11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$204.00 - $482.50

Specifications

Antigen Pepsinogen C/PGC/Progastricsin
Clone 90/11
Concentration 1.0 mg/mL
Dilution ELISA 1:100-1:2000
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017515
SDP
View Documents
Novus Biologicals
NB100628190.025MG
0.025 mg
Each for $204.00
Add to Cart
 
NB10062819
SDP
View Documents
Novus Biologicals
NB10062819
0.1 mg
Each for $482.50
Add to Cart
 
Description

Description

Pepsinogen C/PGC/Progastricsin Monoclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.
Specifications

Specifications

Pepsinogen C/PGC/Progastricsin
1.0 mg/mL
ELISA
Unconjugated
RUO
EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C)
PGA3
IgG1
Affinity Purified
NB100-62819 is specific for human pepsinogen II. Less than 1% reactivity has been observed with human pepsinogen I (isozymogens 2-5), porcine pepsinogen, gastrin I, somatostatin, peptide YY and bombesin. Affinity: 1 x 109 l/mol
90/11
ELISA 1:100-1:2000
Monoclonal
Mouse
Human
643834
Human pepsinogen II
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit