Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen I Mouse anti-Human, Clone: 8003 (99/12), Novus Biologicals™
Mouse Monoclonal Antibody
$176.85 - $436.00
Specifications
Antigen | Pepsinogen I |
---|---|
Clone | 8003 (99/12) |
Concentration | 0.5 mg/mL |
Dilution | ELISA 1:100-1:2000, Immunohistochemistry-Paraffin, Radioimmunoassay 1:100-1:2000 |
Applications | ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10062239
|
Novus Biologicals
NB10062239 |
0.1 mg |
Each for $436.00
|
|
NB017448
|
Novus Biologicals
NB100622390.025MG |
0.025 mg |
Each for $176.85
|
|
Description
Pepsinogen I Monoclonal specifically detects Pepsinogen I in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Radioimmunoassay.Specifications
Pepsinogen I | |
0.5 mg/mL | |
ELISA, Immunohistochemistry (Paraffin), Radioimmune Assays | |
Unconjugated | |
Mouse | |
Human | |
643834 | |
Purified human pepsinogen 1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
8003 (99/12) | |
ELISA 1:100-1:2000, Immunohistochemistry-Paraffin, Radioimmunoassay 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
pepsinogen 3, group I (pepsinogen A) | |
PGA3 | |
IgG1 | |
Protein A purified | |
NB100-62239 recognises human Pepsinogen I, a zymogen or proenzyme secreted by Chief cells in the stomach. It is cleaved to form pepsin both in an autocatalytic fashion and by pepsin itself. In humans there are two related forms of pepsin, Pepsinogen I (also known as pepsinogen A), and Pepsinogen II (also known as Pepsinogen B or progastricsin). |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title