Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pepstatin A

Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D.

$45.40 - $149.00

Specifications

Synonym Isovaleryl-val-val-4-amino-3-hydroxy-6-methylheptanoyl-ala-4-amino-3-hydroxy-6-methylheptanoic acid SDS Certificate of Analysis Product Specification Technical Inquiry
Format Powder
Molecular Formula C34H63N5O9
CAS 26305-03-3
Concentration 10-6 to 10-9M
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ20037LB0 View Documents Affymetrix/USB™
J20037LB0
5mg Each for $45.40
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ20037MB View Documents Thermo Scientific™
J20037MB
25mg Each for $149.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • Effective concentrations are 10-6 to 10-9M
  • Does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin
  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • MDL: MFCD00060740
Specifications

Specifications

Isovaleryl-val-val-4-amino-3-hydroxy-6-methylheptanoyl-ala-4-amino-3-hydroxy-6-methylheptanoic acid SDS Certificate of Analysis Product Specification Technical Inquiry
C34H63N5O9
10-6 to 10-9M
Powder
26305-03-3
Dissolves in methanol
Safety and Handling

Safety and Handling

EINECSNumber : 247-600-0

Recommended Storage : Keep cold

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit