Learn More
Affymetrix/USB™ Pepstatin A
Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D.
Manufacturer:  Affymetrix/USB™ J20037LB0
 View more versions of this product
Catalog No. AAJ20037LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • Effective concentrations are 10-6 to 10-9M
  • Does not inhibit Trypsin, Chymotrypsin, Papain or Plasmin
  • Protease inhibitor, inhibits Pepsin, Renin and Cathepsin D
  • MDL: MFCD00060740
Specifications
Specifications
10-6 to 10-9M
5mg
C34H63N5O9
Dissolves in methanol
Powder
26305-03-3
Isovaleryl-val-val-4-amino-3-hydroxy-6-methylheptanoyl-ala-4-amino-3-hydroxy-6-methylheptanoic acid SDS Certificate of Analysis Product Specification Technical Inquiry