Learn More
Thermo Scientific™ PepSwift™ Monolithic Capillary HPLC Columns Available on GSA/VA Contract for Federal Government customers only.
Use for fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications.
$1044.00 - $1644.00
Specifications
Column Type Reversed Phase
Packing Material Monolith Polymer
pH 1 to 10
Pore Size Monolith
Stationary Phase PSDVB (Polystyrene Divinylbenzene)
View More Specs
Products 4
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
DX164558 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
164558
Monolith 0.2 mm 5 mm
Pack of 2 for $1,044.00
Add to cart
DX164557 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
164557
Monolith 0.2 mm 50 mm
Each for $1,308.00
Add to cart
DX164543
View Documents
Thermo Scientific™
164543
Monolith 0.1 mm 250 mm
Each for $1,644.00
Add to cart
DX164585
View Documents
Thermo Scientific™
164585
Monolith 0.5 mm 50 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
For fast, high-resolution LC/MS separations in bottom-up protein identification, biomarker discovery, and systems biology applications, use Thermo Scientific™ PepSwift™ Monolithic Capillary LC Columns. Based on a polystyrene divinylbenzene copolymer, the monolithic structure offers a high-quality alternative to traditional microparticulate sorbents, providing important advantages to the chromatographic separation. High-sensitivity proteomics and biotech applications are easily performed using these columns.
Benefit from:
  • High-speed peptide and protein separations (<15 min)
  • High sensitivity for LC/MS
  • High column-to-column reproducibility
  • Wide range of column I.D. and lengths available
  • Superior lifetime
  • Easy column installation using Thermo Scientific™ Dionex™ nanoViper™ Fingertight Fittings
  • Precolumns available to preconcentrate and desalt samples
You can also use Thermo Scientific™ PepSwift™ Trap Columns for preconcentration and desalting of samples consisting of peptides and proteins without negatively impacting the chromatographic performance or recovery of the compounds. Change the selectivity of the separation or improve the trapping efficiency by employing various ion-pairing agents in the loading solvent and/or mobile phases. The trap columns are used with Thermo Scientific™ PepSwift™ and ProSwift™ Monolithic Columns, which are for fast, high-resolution LC/MS separations in protein identification, biomarker discovery, and systems biology applications.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Reversed Phase
1 to 10
PSDVB (Polystyrene Divinylbenzene)
Monolith Polymer
Monolith
SDS
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.