Learn More
Bachem Peptide YY (3-36) (canine, mouse, porcine, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4044052.0500
Encompass_Preferred
Peptide YY (3-36) (canine, mouse, porcine, rat) Trifluoroacetate, H-Ala-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Ser-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2, 126339-09-1, C176H272N52O54, Mr 3980.41, 0.5mg. PYY (3-36) (canine, mouse, porcine, rat). Peptide YY (3-36) and peptide YY are both synthesized by the gastrointestinal tract and released into the circulation after a meal. Peptide YY (3-36) is a Y2 receptor subtype agonist, whereas peptide YY is non-selective for Y1 and Y2 receptor subtypes. It has been suggested that Y1 and Y2 receptor subtype binding affinities depend on their secondary and tertiary solution state structures.
Catalog No. 50-194-6924
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.