Learn More
Echelon Biosciences Research Labs PYY / Peptide YY (3-36), human - 1 mg, CAS#: 123583-37-9
SDP
Supplier:  Echelon Biosciences Research Labs 490111MG
Encompass_Preferred
PYY / Peptide YY (3-36), human - 1mg , Ile-Lys-Pro-Glu-Ala-Pro-Gly-Glu-Asp-Ala-Ser-Pro-Glu-Glu-Leu-Asn-Arg-Tyr-Tyr-Ala-Ser-Leu-Arg-His-Tyr-Leu-Asn-Leu-Val-Thr-Arg-Gln-Arg-Tyr-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8544
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.