Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peptide YY Mouse anti-Human, Alexa Fluor™ 594, Clone: 4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223584AF594

Catalog No. NP22384A94


Add to cart

Description

Description

Peptide YY Monoclonal antibody specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Peptide YY
4
Western Blot, ELISA
Conjugated, Purified
PYY
Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier
Protein A or G purified
Signal Transduction
Primary
5697.0
Human
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Clone 4 binds human PYY(3-36) Based on inhibition ELISA, Clone 4 shows no cross-reaction with PYY(1-36), PYY from mouse, rat, dog or pig, or NPY.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit