Learn More
PeptideYY Mouse anti-Human, Janelia Fluor 646, Clone: 1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP105167JF646
Catalog No. NBP15167646
Add to cart
Description
Description
PeptideYY Monoclonal antibody specifically detects PeptideYY in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
PeptideYY
1
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
PYY
Synthetic human PYY adsorbed onto aluminum hydroxide gel
Protein A or G purified
Signal Transduction
Primary
5697
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I
Mouse
IgG1 Kappa
0.1mL
Store at 4°C in the dark.
Monoclonal
Binds human PYY. Cross-reactivity with NPY or PP in solution is