Learn More
Anaerobe SystemsSupplier Diversity Partner PY RIBOSE - Peptone Yeast Extract Broth with Ribose, 10 Tubes/Pkg (NON-STOCK, 10 PACK MINIMUM ORDER) DFS Item
Manufacturer:  Anaerobe SystemsSupplier Diversity Partner AS854
Peptone Yeast Extract Broth Based Media are enriched nonselective broth used for growth & identification of anaerobic bacteria based on their biochemical features. This media is prepared, dispensed, and packaged under oxygen-free conditions to prevent the formation of oxidized products prior to use.PY Broth contains hemin & vitamin K for the recovery of fastidious organisms such as Prevotella spp and Porphyromonas spp. PRAS (Pre-reduced anaerobically sterilized) biochemicals are considered the gold standard for biochemical testing and characterization of anaerobes. Adding specific biochemical, the Peptone Yeast Extract Broth Based Media can determine whether anaerobic bacteria will utilize the biochemical or if growth is stimulated or inhibited. PY was originally formulated for use in chromatographic analysis of metabolic products from the fermentation of glucose. This chromatographic analysis is useful in the identification of clinically important anaerobic bacteria.
Catalog No. NC1474167