Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PER2 Mouse anti-Human, DyLight 650, Clone: 5310, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243729C

Catalog No. NBP243729C

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PER2
5310
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PER2
Recombinant protein encompassing a sequence within the N-terminus region of human PER2. The exact sequence is proprietary.
Protein G purified
Cancer, Chromatin Research, Circadian Rhythm, Endothelial Cell Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
Primary
8864
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Circadian clock protein PERIOD 2, FASPS, hPER2, KIAA0347period 2, period (Drosophila) homolog 2, period circadian protein 2, period circadian protein homolog 2, period homolog 2 (Drosophila)
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit