Learn More
PER2 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 203591AP
Catalog No. 50-173-2846
Add to cart
Description
Description
The antibody is specific to PER2. It can recognize isoform 1 and isoform 2 of PER2.
PER2, also known as Period circadian protein homolog 2, is a mainly nuclear protein that shows nucleocytoplasmic shuttling which is effected by interaction with other circadian core oscillator proteins and/or by phosphorylation. PER2 belongs to the basic helix-loop-helix family of transcription factors and contains a PAC (PAS-associated C-terminal) domain and two PAS (PER-ARNT-SIM) domains. PER2 is a component of the circadian core oscillator, which includes the CRY proteins, CLOCK or NPAS2, BMAL1 or BMAL2, CSNK1D and/or CSNK1E, TIMELESS, and the PER proteins. This protein is thus essential for generating circadian rhythms and is a negative element in the circadian transcriptional loop which influences clock function by interacting with other circadian regulatory proteins and transporting them to the nucleus. Expression of PER2 is fairly wide spread with strong expression in skeletal muscle and pancreas and slight expression in lung. Defects in PER2 are a cause of familial advanced sleep-phase syndrome (FASPS).
Specifications
Specifications
PER2
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15055
FASPS, hPER2, KIAA0347, PER2, period homolog 2 (Drosophila)
Rabbit
IgG
150 μL
-20°C
Primary
8864
Immunoprecipitation, Western Blot
Unconjugated
PER2
Liquid
PER2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human