Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PER2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | PER2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB024128
|
Novus Biologicals
NBP294809100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB024129
|
Novus Biologicals
NBP29480920UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
PER2 Polyclonal antibody specifically detects PER2 in Mouse samples. It is validated for Western BlotSpecifications
PER2 | |
Polyclonal | |
Purified | |
RUO | |
Mouse | |
Circadian clock protein PERIOD 2, FASPS, hPER2, KIAA0347period 2, period (Drosophila) homolog 2, period circadian protein 2, period circadian protein homolog 2, period homolog 2 (Drosophila) | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PER2 (NP_073728.1). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cancer, Chromatin Research, Circadian Rhythm, Endothelial Cell Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision | |
PBS (pH 7.3), 50% glycerol | |
8864 | |
IgG | |
Affinity purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title