Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PER3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540922
Description
Peptide sequence: DATLFCEPWT LNMQPAPLTS EEFKHVGLTA AVLSAHTQKE EQNYVDKFRE Sequence homology: Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 86%; Rat: 93%; Sheep: 86%; Zebrafish: 79%.
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. This gene is upregulated by CLOCK/ARNTL heterodimers but then represses this upregulation in a feedback loop using PER/CRY heterodimers to interact with CLOCK/ARNTL. Polymorphisms in this gene have been linked to sleep disorders. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
PER3 | |
Polyclonal | |
Unconjugated | |
PER3 | |
2810049O06Rik; Cell growth-inhibiting gene 13 protein; circadian clock protein PERIOD 3; FASPS3; GIG13; hPER3; mPER3; Per3; period circadian clock 3; period circadian protein 3; period circadian protein homolog 3; period homolog 3; period3; rper3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
8863 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P56645 | |
PER3 | |
Synthetic peptide directed towards the C-terminal of human PER3 (aa 647-696). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction