Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PER3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | PER3 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PER3 Polyclonal specifically detects PER3 in Human samples. It is validated for Western Blot.Specifications
PER3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Cell growth-inhibiting gene 13 protein, Circadian clock protein PERIOD 3, GIG13, growth-inhibiting protein 13, hPER3, period (Drosophila) homolog 3, period 3, period circadian protein 3, period circadian protein homolog 3, period homolog 3 (Drosophila) | |
The immunogen is a synthetic peptide directed towards the N terminal region of human PER3 (NP_058515). Peptide sequence GAPQADVSMYSLEELATIASEHTSKNTDTFVAVFSFLSGRLVHISEQAAL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
8863 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title