Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PER3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310332100UL
Description
PER3 Polyclonal specifically detects PER3 in Human samples. It is validated for Western Blot.Specifications
PER3 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Cell growth-inhibiting gene 13 protein, Circadian clock protein PERIOD 3, GIG13, growth-inhibiting protein 13, hPER3, period (Drosophila) homolog 3, period 3, period circadian protein 3, period circadian protein homolog 3, period homolog 3 (Drosophila) | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PER3. Peptide sequence DATLFCEPWTLNMQPAPLTSEEFKHVGLTAAVLSAHTQKEEQNYVDKFRE | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
8863 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction