Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PER3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen PER3
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125565
View Documents
Novus Biologicals
NBP310332100UL
100 μg
Each for $320.71
N/A
NB125566
View Documents
Novus Biologicals
NBP31033225UL
25 μg
Each for $106.19
N/A
Description

Description

PER3 Polyclonal antibody specifically detects PER3 in Human samples. It is validated for Western Blot
Specifications

Specifications

PER3
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
8863
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
Cell growth-inhibiting gene 13 protein, Circadian clock protein PERIOD 3, GIG13, growth-inhibiting protein 13, hPER3, period (Drosophila) homolog 3, period 3, period circadian protein 3, period circadian protein homolog 3, period homolog 3 (Drosophila)
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PER3. Peptide sequence DATLFCEPWTLNMQPAPLTSEEFKHVGLTAAVLSAHTQKEEQNYVDKFRE
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit