Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Pierce™ Staphylococcus aureus V-8 Protease and Kt

Specifically cleave only at glutamic acid residues or cleave at both glutamic and aspartic residues with our immobilized Staphylococcus aureus V-8 protease kit.

$728.00

Specifications

MS-Compatible No
Storage Requirements 4°C
View More Specs

Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
PI20151
View Documents
Thermo Scientific™
20151
Pierce Staphylococcus aureus V-8 Protease Kit
Pack for $728.00
Add to cart
 
Description

Description

Thermo Scientific™ Pierce V-8 Protease Kit contains Staphylococcus aureus V-8 protease immobilized on resin and three digestion buffers for specific cleavage at glutamic acid residues or cleavage at glutamic and aspartic acid residues.

Has two pH optima, one at pH 4.0 and the other at pH 7.8 and is specific for cleavage at the carboxy terminus of glutamic acid and aspartic acid residues. The protease is further specific for only glutamic digestion in buffers, which do not contain phosphate at either pH optimum. Phosphate buffers at pH 7.8 will facilitate digestion at both glutamic and aspartic acid residues.

Highlights:

  • Specificity for glutamic acid is achieved in ammonium bicarbonate, pH 7.8 or ammonium acetate, pH 4
  • Enzyme is active in the presence of many denaturing agents such as SDS, urea and guanidine·HCl
Specifications

Specifications

No
4°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit