Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.7, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Poly Bottle
7601-90-3
ClHO4
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2,perchloric acid,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchioric acid,ueberchlorsaeure,hyperchloric acid,hclo4
24247
CHEBI:29221
perchloric acid
OCl(=O)(=O)=O
7601-90-3
ClHO4
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2,perchloric acid,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchioric acid,ueberchlorsaeure,hyperchloric acid,hclo4
24247
CHEBI:29221
perchloric acid
OCl(=O)(=O)=O

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use
Quantity 2.5L
Formula Weight 100.46
Percent Purity 70%
Grade BAKER INSTRA-ANALYZED™ Plus
Packaging Poly Bottle
Specific Gravity 1L = 1.68kg
Chemical Name or Material Perchloric Acid