Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 60 to 62%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
PVC Coated Glass Bottle

Chemical Identifiers

7601-90-3
ClHO4
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
CHEBI:29221
perchloric acid
OCl(=O)(=O)=O

High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Formula Weight 100.46
Packaging PVC Coated Glass Bottle
Specific Gravity 1L = 1.50kg
Chemical Name or Material Perchloric Acid
Assay 60 to 62%
Grade Baker Analyzed™ ACS Reagent