Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 69 to 72%, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
PVC Coated Glass Bottle

Chemical Identifiers

7601-90-3
ClHO4
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
CHEBI:29221

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    For Trace Metal Analysis
Quantity 500mL
SMILES OCl(=O)(=O)=O
IUPAC Name perchloric acid
Formula Weight 100.46
Packaging PVC Coated Glass Bottle
Specific Gravity 1L = 1.50kg
Chemical Name or Material Perchloric Acid
Assay 69 to 72%
Grade BAKER INSTRA-ANALYZED™ Reagent