Perchloric acid, ACS, 48-50%, Alfa Aesar™

Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm

Manufacturer: Alfa Aesar 44464AE

 View more versions of this product

Catalog No. AA44464AE

  Check Availability
Add to Cart

    Specifications

    Purity Grade ACS Grade
    Formula Weight 100.46
    Quantity 100mL
    DOT Information DOT Class: 8, Packaging Group: II
    Health Hazard 2 Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
    Density 1.41g/cm3 at 20°C
    EINECS Number 231-512-4
    Physical Form Liquid
    UN Number UN1802
    Chemical Name or Material Perchloric acid
    Vapor Pressure 29mm Hg at 20°C
    Assay Percent Range 48-50%
    Molecular Formula HClO4
    MDL Number MFCD00011325
    CAS 7601-90-3
    Health Hazard 1 H272-H314: May intensify fire; oxidizer. Causes severe skin burns and eye damage.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit