Learn More
Perchloric Acid, ACS, 60-62%, Thermo Scientific Chemicals
Description
Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals, alkali metals such as sodium and potassium. Also, used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 12°C |
Boiling Point | 203°C |
Density | 1.54 |
Flash Point | 113°C (235°F) |
Quantity | 100 mL |
Physical Form | Liquid |
Solubility Information | Miscible with water. |
Formula Weight | 100.46 |
Grade | ACS Reagent |
Chemical Name or Material | Perchloric Acid |
Safety and Handling
Hazard Category | H271-H290-H302-H314-H335-H373 |
Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: I |
EINECSNumber | 231-512-4 |
RTECSNumber | SC7500000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.