Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric acid, ACS, 69.0-72.0%, redistilled, Thermo Scientific Chemicals

Catalog Number aa87963ap
Click to view available options
Quantity:
500 mL
2.5 L
7601-90-3
ClHO4
100.45
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
24247
CHEBI:29221
perchloric acid
O[Cl](=O)(=O)=O
7601-90-3
ClHO4
100.45
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
24247
CHEBI:29221
perchloric acid
O[Cl](=O)(=O)=O

Perchloric acid is used as a precursor for ammonium perchlorate which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -112°C
Boiling Point 203°C
Density 1.67 g/mL
Flash Point 113°C (235°F)
Assay Percent Range 70%
Quantity 500 mL
Physical Form Liquid
UN Number UN1873
Merck Index 14,7153
Solubility Information Miscible with water.
Formula Weight 100.46
Percent Purity 70%
Grade ACS
Chemical Name or Material Perchloric Acid, redistilled, Redistilled
Show More Show Less
Hazard Category H271-H290-H302-H314-H335-H373
Hazard Statement GHS H Statement
H271-H314
Precautionary Statement P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID
EINECSNumber 231-512-4
RTECSNumber SC7500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit