Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric acid concentrate, For 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium, Honeywell Fluka™

Catalog Number p-7122056
Click to view available options
Quantity:
1 L
6 x 1 Ea.
Packaging:
Ampoule
Bottle
7601-90-3
ClHO4
100.45
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
24247
CHEBI:29221
perchloric acid
O[Cl](=O)(=O)=O
7601-90-3
ClHO4
100.45
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
24247
CHEBI:29221
perchloric acid
O[Cl](=O)(=O)=O

for 1L standard solution, 0.1 M HClO4 (0.1N), for titration in non-aqueous medium

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Packaging Bottle
Flash Point 113°C (235.4°F)
Quantity 1 L
Linear Formula HClO4
UN Number 1873
Formula Weight 100.46g/mol
Chemical Name or Material Perchloric Acid Concentrate
Hazard Statement P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375
Precautionary Statement H271-H290-H302-H314-H373
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit