Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Perchloric acid, Environmental Grade, 67-71%

Manufacturer:  Alfa Aesar™ 38749AP

Quantity 500mL

 View more versions of this product

Catalog No. AA38749AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7601-90-3
MFCD00011325
14,7153
Environmental Grade
100.46
UN1873
Perchloric acid

Specifications

Colorless
7601-90-3
100.46
UN1873
1.67g/mL
113° (235°F)
203°C
65 to 71%
500mL
Liquid, Single Sub-Boiling Quartz Distillation
MFCD00011325
14,7153
Odorless
−12°C
Perchloric acid
Environmental Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement
H271-H314
May cause fire or explosion
strong oxidizer.
Causes severe skin burns and eye damage.

GHS P Statement
P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID

RTECSNumber : SC7500000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit