Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, Normality about 12, 69 to 72%, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass Bottle
PVC Coated Glass Bottle

Chemical Identifiers

7601-90-3
ClHO4
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
24247
CHEBI:29221

High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
SMILES OCl(=O)(=O)=O
IUPAC Name perchloric acid
Formula Weight 100.46
Packaging PVC Coated Glass Bottle
Specific Gravity 1L = 1.67kg
Chemical Name or Material Perchloric Acid, Normality about 12
Assay 69 to 72%
Grade Baker Analyzed™ ACS Reagent