Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric acid, pure, 0.1M solution in acetic acid, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
7601-90-3,64-19-7
ClHO4
100.454
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchioric acid,ueberchlorsaeure,hyperchloric acid,hclo4
24247
CHEBI:29221
perchloric acid
OCl(=O)(=O)=O
7601-90-3,64-19-7
ClHO4
100.454
MFCD00011325
VLTRZXGMWDSKGL-UHFFFAOYSA-N
unii-v561v90bg2,perchloric acid,perchloric acid acid by mass forbidden,perchloric acid, acs reagent,perchlorsaeure,perchioric acid,ueberchlorsaeure,hyperchloric acid,hclo4
24247
CHEBI:29221
perchloric acid
OCl(=O)(=O)=O
Oxidation reagent
Color Colorless
Quantity 1L
UN Number 2920
Fieser 01,796; 02,309; 03,220; 06,453; 09,279; 11,402
Merck Index 15, 7267
Solubility Information Solubility in water: insoluble.
Density 1.0600g/mL
Formula Weight 100.46
Physical Form Liquid
Grade Pure
Factor 0.998 to 1.002
Packaging Glass bottle
Solubility Solubility in water: insoluble.
Specific Gravity 1.06
Water 0.2% max. (K.F.)
Flash Point 40°C
Chemical Name or Material Perchloric acid, 0.1 M Solution in Acetic Acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger