Learn More
BiolegendSupplier Diversity Partner PerCP anti-human CD4  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 344624
PerCP anti-human CD4 [SK3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-712-090