Learn More
BiolegendSupplier Diversity Partner PerCP/Cy5.5 anti-human CD107a (LAMP-1) DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 328615
PerCP/Cy5.5 anti-human CD107a (LAMP-1) [H4A3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Pigtailed Macaque, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-461