Learn More
BiolegendSupplier Diversity Partner PerCP/Cyanine5.5 anti-mouse CD172a (SIRPα)  DFS Item
Manufacturer:  BiolegendSupplier Diversity Partner 144010
PerCP/Cyanine5.5 anti-mouse CD172a (SIRPα) [P84]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-711-893