Learn More
Honeywell™ Perfect Fit™ Aramid Fiber Dotted Gloves
Exceptional cut, slash and flame resistance
Manufacturer:  Honeywell™ KVD18AR100
 View more versions of this product
Catalog No. 19-050-547
Add to cart
Description
Description
Seamless knit glove made of 100% Kevlar™ Aramid fibers brand fiber. Kevlar™ Aramid fibers is, pound-for-pound, five times stronger than steel, providing exceptional protection against cut, slash and abrasion. Kevlar™ Aramid fibers fiber will not support a flame and can withstand temperatures as high as 480°C (900°F) without breaking down. This glove is not recommended for thermal heat protection.
  • Extremely durable; seamless construction eliminates split seams and is more comfortable than gloves with seams that chafe
  • Ambidextrous glove has rugged, high-quality brown dot coating on both sides to enhance grip and abrasion resistance
  • Knitwrist
  • ANSI™ cut level 3
Specifications
Specifications
Safety
Ambidextrous
Dupont™ Kevlar™ Aramid Fiber
Medium weight
Knit
automotive and part manufacturer, building and construction, fishing, general industry, Glass industries, Metal Steel, mining and quarrying, ship building, Wood industries
Men's